Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 281, Tocris Bioscience™

Catalog No. p-4817006 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

STK33 inhibitor

Inhibitor of the serine/threonine kinase STK33 (IC50 = 14 nM). Exhibits >700-fold selectivity for STK33 over the structurally related protein kinase A (PKA); also exhibits 550-fold selectivity over Aurora kinase B.

Chemical Identifiers

CAS 1404437-62-2
Molecular Formula C22H19N3O2S
Molecular Weight (g/mol) 389.473
InChI Key HWOYIOLMBQSTQS-UHFFFAOYSA-N
Synonym bg-stk33-59, n-2-3,4-dihydro-3-oxo-2-quinoxalinyl-4-1-methylethyl phenyl-2-thiophenecarboxamide, n-2-3-oxo-3,4-dihydroquinoxalin-2-yl-4-propan-2-yl phenyl thiophene-2-carboxamide
PubChem CID 53377448
IUPAC Name N-[2-(3-oxo-4H-quinoxalin-2-yl)-4-propan-2-ylphenyl]thiophene-2-carboxamide
SMILES CC(C)C1=CC(=C(C=C1)NC(=O)C2=CC=CS2)C3=NC4=CC=CC=C4NC3=O

Specifications

Quantity 10 mg
Formula Weight 389.47
Percent Purity >98%
Chemical Name or Material ML 281
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.