Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 7 hydrochloride, Tocris Bioscience™

Catalog No. p-4817086 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of myosin light chain kinase

Selective MLCK inhibitor (Ki = 0.3 μM). Exhibits more potent inhibition than ML 9 hydrochloride (Cat. No. 0431) Displays reversible, ATP-competitive inhibition of Ca2+-calmodulin-dependent and -independent smooth muscle MLCKs. Inhibits proplatelet formation and stabilization.

Chemical Identifiers

CAS 110448-33-4
Molecular Formula C15H18ClIN2O2S
Molecular Weight (g/mol) 452.735
InChI Key KDDALCDYHZIZMH-UHFFFAOYSA-N
Synonym ml-7 hydrochloride, ml-7, 1-5-iodonaphthalen-1-yl sulfonyl-1,4-diazepane hydrochloride, ml 7 hydrochloride, ml-7, hydrochloride, ml-7 hcl, 1-5-iodonaphthalene-1-sulfonyl-1h-hexahydro-1,4-diazepine hydrochloride, 1-5-iodonaphthalen-1-ylsulfonyl-1,4-diazepane hydrochloride, 1-5-iodonaphthalene-1-sulfonyl homopiperazine, hcl
PubChem CID 9803932
IUPAC Name 1-(5-iodonaphthalen-1-yl)sulfonyl-1,4-diazepane;hydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CC=C3I.Cl

Specifications

Quantity 10 mg
Formula Weight 452.74
Percent Purity 0.98
Chemical Name or Material ML 7 hydrochloride
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.