Learn More
Medchemexpress LLC HY-12345 5mg Medchemexpress, ML365 CAS:947914-18-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123455MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-12345 5mg ML365 CAS:947914-18-3 ML365 is a selective inhibitor of KCNK3/TASK1 two-pore domain potassium channel, with an IC50 of 4 nM (thallium flux assay). ML365 acts as a pharmacological tool that can be used to examine the specific roles of TASK1 channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1017
missing translation for 'QuantityLeft'