Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
MLS 1547 is a G protein-biased D2 receptor partial agonist (Ki = 1.2 μM). Stimulates D2 receptor G protein-mediated signaling and acts as an antagonist of dopamine-mediated β-arrestin recruitment at D2 receptors.
Specifications
Specifications
Chemical Name or Material | 5-Chloro-7-[[4-(2-pyridinyl)-1-piperazinyl]methyl]-8-quinolinol |
CAS | 315698-36-3 |
Assay Percent Range | >98% |
Quantity | 25 mg |
Molecular Formula | C19H19ClN4O |
Formula Weight | Observed MW: 354.83 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction