Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MM 11253, Tocris Bioscience™

Catalog No. p-4817400 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

RARγ-selective antagonist

Selective RARγ antagonist. Blocks the growth inhibitory ability of RARγ-selective agonists in squamous cell carcinoma (SCC)-25 cells.

Chemical Identifiers

CAS 345952-44-5
Molecular Formula C28H30O2S2
Molecular Weight (g/mol) 462.666
InChI Key DPVGXUNWCVEIGT-UHFFFAOYSA-N
Synonym d06ram, 6-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl-1,3-dithiolan-2-yl naphthalene-2-carboxylic acid, 6-2-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl-1,3-dithiolan-2-yl-2-naphthalenecarboxylic acid
PubChem CID 10389639
IUPAC Name 6-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)-1,3-dithiolan-2-yl]naphthalene-2-carboxylic acid
SMILES CC1(CCC(C2=C1C=CC(=C2)C3(SCCS3)C4=CC5=C(C=C4)C=C(C=C5)C(=O)O)(C)C)C

Specifications

Quantity 10 mg
Formula Weight 462.67
Percent Purity >98%
Chemical Name or Material MM 11253
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.