Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moclobemide, Tocris Bioscience™

Catalog No. p-4817253 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reversible MAO-A inhibitor

Reversible monoamine oxidase A (MOA-A) inhibitor.

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13H17ClN2O2
Molecular Weight (g/mol) 268.741
InChI Key YHXISWVBGDMDLQ-UHFFFAOYSA-N
Synonym moclobemide, aurorix, manerix, moclamine, moclobemid, moclobemida, moclobemidum, moclaime, moclamide, moclobamide
PubChem CID 4235
ChEBI CHEBI:83531
IUPAC Name 4-chloro-N-(2-morpholin-4-ylethyl)benzamide
SMILES C1COCCN1CCNC(=O)C2=CC=C(C=C2)Cl

Specifications

Quantity 10 mg
Formula Weight 268.74
Percent Purity >99%
Chemical Name or Material Moclobemide
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.