Learn More
MilliporeSigma™ Supelco™ Molded Thermogreen™ LB-2 Septa with Injection Hole
Molded Septa are manufactured from high quality, low bleed material that uses the same exclusive LB-2 rubber formulation chromatographers are accustomed to using.
Supplier:  MilliporeSigma™ Supelco™ 28331U
 View more versions of this product
Catalog No. 11-101-0650
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Molded Septa offer easier installation and better sealing inside the injection port
  • A liquid injection molding process ensures that every septum conforms to the same shape with crisp, clean sides
  • Injection hole helps guide the syringe needle to puncture the same location every injection
  • Benefits of the injection hole include minimal coring and less septum fragments that could contaminate the inlet liner
  • High puncture tolerance makes these septa ideal for use with autosampler injections, manual injections, and/or SPME applications
  • The useable inlet temperature range of 100-350°C is adequate for most GC applications
  • Rubber formulation is exclusive to Supelco
  • Strict tolerances (diameter, thickness, injection hole) due to constant dimension of the mold itself
  • Ultra low bleed over a wide range of inlet temperatures
  • No foreign substances/powders, which could contaminate the inlet, are used during manufacturing
  • Fully tested for bleed and contamination
  • Pre-conditioned and ready to use
  • Ideal for use with low bleed GC-MS columns
Specifications
Specifications
350°C
LB-2 Rubber
Septa
50 Pk.
9.5 mm
Low Bleed GC-MS Columns
With Injection Hole
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC