Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ Mellows™ Disposable Ear Plugs
SDP

Mellows™ disposable ear plugs are made of extra-soft, extra-light foam and are appropriate for mid- to high-level noise areas.

$16.66

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
High Visibility Yes
View More Specs

Products 2
Catalog Number Mfr. No. Corded Price Quantity & Availability  
Catalog Number Mfr. No. Corded Price Quantity & Availability  
19-384-342
SDP
View Documents
Moldex™
6840
Corded
Pack of 100 for $16.66

Save 72%

Reg : $60.30

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Case of 20 PK for $333.17

Save 71%

Reg : $1,145.79

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
missing translation for 'QuantityLeft'
Add to Cart
 
19-384-341
SDP
View Documents
Moldex™
6820
Uncorded N/A N/A N/A
Description

Description

  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Non-irritating smooth surface
  • High visibility color makes compliance checks quick and easy
  • Shaped to fit even very small ear canals
Specifications

Specifications

Yes
Universal
Yes
Foam
30 dB
Single-use
Class AL
Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
Poly bag
Orange
Bullet
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.