Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 594 conjugate
SDP

Catalog No. A13101
Encompass
Click to view available options
:
5mg

Bright and photostable conjugate exceptionally suited for applications in which well-defined molecular weight tracers are needed

  • Applications include studying rate of endocytosis and exocytosis and following intracellular protein processing

Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 594
Species Bovine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins
Product Line Alexa Fluor™

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.