Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lectin PHA-L From Phaseolus vulgaris (red kidney bean), Alexa Fluor™ 488 Conjugate
SDP

Catalog No. L11270
Encompass
Click to view available options
Quantity:
1 mg

Suitable for use in cell tracing and tracking, cellular imaging and flow cytometry

Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues.[ALTREF PN29369] This lectin has been shown to agglutinate leukocytes and induce mitogenic activity in T-lymphocytes[ALTREF PN29369,PN29368] and has also been used as an anterograde tracer for the study of neuronal pathways.[ALTREF PN20284]

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence Staining & Detection, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 mg
Source Red kidney bean
Conjugate Alexa Fluor 488
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor™

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.