Learn More
Invitrogen™ PeppermintStick™ Phosphoprotein Molecular Weight Standards

Molecular weight range from 14,400 to 116,250
$303.00 - $386.00
Description
PeppermintStick™ phosphoprotein molecular weight standards are a mixture of phosphorylated and nonphosphorylated proteins. Separation by polyacrylamide gel electrophoresis resolves the standard mixture into two phosphorylated and four nonphosphorylated protein bands.
- System: SDS-PAGE
- Stain Type: Unstained
- Ready to Load: No
- Gel Compatibility: NuPAGE™ Tris-Acetate Gels, Novex™Tricine Gels, NuPAGE™ Bis-Tris Gels, Novex™Tris-Glycine Gels, SDS-PAGE Gels
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.