Learn More
Ampco Safety Tools Monkey Wrench
Shop All Ampco Metal Inc. Products

Certifications
FM Approved
Warranty and Services
Lifetime Warranty with proper use. If a tool breaks or fails to perform under normal use aor service, it will be repaired or eplaced at no charge - exceptions include defects resulting from misuse, abuse or normal service wear.
Specifications
For Use With (Equipment)
Electronics, batteries, MRI, and precision magnetic equipment
Length (English)
8 in.
Length (Metric)
203.2 mm
Material
Aluminum Bronze
Color
Bronze
For Use With (Application)
Non-Sparking, Non-Magnetic, Corrosion Resistant Applications; Flammable or combustible vapors, liquids, dusts or residues; Corrosive environments like laboratories, acid, salt water and chemical processing
No. per Pack
1 Ea.
Weight (Metric)
635 g
Width (Metric)
53.97 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.