Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MONNA, >98%, Tocris Bioscience™

Catalog No. p-7121263 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent TMEM16A (Anoctamin-1) blocker

MONNA is a potent TMEM16A (Anoctamin-1) blocker (IC50 = 80 nM). Exhibits >100-fold selectivity for TMEM16A over bestrophin-1, CLC2 and CFTR. Induces vasorelaxation of rodent resistance arteries in presence or absence of chloride ions. Carbachol (Cat. No. 2810) induced contraction of bronchial myocytes is reduced by MONNA and intracellular calcium release is increased.

Chemical Identifiers

CAS 1572936-83-4
Molecular Formula C18H14N2O5
Molecular Weight (g/mol) 338.319
InChI Key JIVRLHBAUUZTNC-UHFFFAOYSA-N
Synonym monna, 2-4-methoxynaphthalen-2-yl amino-5-nitrobenzoic acid, 2-4-methoxynaphthalen-2-yl amino-5-nitrobenzoic, n-4-methoxy-2-naphthyl-5-nitroanthranilic acid, 2-4-methoxy-2-naphthalenyl amino-5-nitro-benzoic acid
PubChem CID 72165188
IUPAC Name 2-[(4-methoxynaphthalen-2-yl)amino]-5-nitrobenzoic acid
SMILES COC1=CC(=CC2=CC=CC=C21)NC3=C(C=C(C=C3)[N+](=O)[O-])C(=O)O

Specifications

Linear Formula C18H14N2O5
Quantity 10 mg
Solubility Information Soluble to 100mM in DMSO
Formula Weight 338.31
Chemical Name or Material 2-[(4-Methoxy-2-naphthalenyl)amino]-5-nitro-benzoic acid
Grade >98%
Recommended Storage Store at 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.