Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mono-6-O-(p-toluenesulfonyl)-alpha-cyclodextrin 85.0+%, TCI America™
SDP

Catalog No. p-7132789 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 32860-56-3
Molecular Formula C43H66O32S
Molecular Weight (g/mol) 1127.029
InChI Key ARQITQMHQNGIEE-FJFUKXEISA-N
PubChem CID 44629832
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2C3C(C(C(O2)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O3)C(C8O)O)CO)CO)CO)CO)CO)O)O

Specifications

Color White
Quantity 200 mg
Formula Weight 1127.03
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mono-6-O-(p-toluenesulfonyl)-alpha-cyclodextrin
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.