Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mono-Methyl 5-nitroisophthalate, 98%, Thermo Scientific™

Catalog No. AC217750250
Click to view available options
:

Specifications

CAS

1955-46-0

CAS Min %

97.5

CAS Max %

100.0

Infrared Spectrum

Authentic

Assay Percent Range

98%

Packaging

Glass bottle

Molecular Formula

C9H7NO6

Quantity

25g

MDL Number

MFCD00009793

Synonym

3-methoxycarbonyl-5-nitrobenzoic acid, 5-nitroisophthalic acid monomethyl ester, monomethyl 5-nitroisophthalate, methyl 5-nitroisophthalate, mono-methyl 5-nitroisophthalate, mono-methyl-5-nitroisophthalic acid, methyl hydrogen 5-nitroisophthalate, 5-nitroisophthalic acid, monomethyl ester, monomethyl-5-nitroisophthalate, monomethyl 5-nitrobenzene-1,3-dicarboxylate

Solubility Information

Solubility in water: insoluble.

InChI Key

ZCRNIIJXDRYWDU-UHFFFAOYSA-N

SMILES

COC(=O)C1=CC(=CC(=C1)C(=O)O)[N+](=O)[O-]

Molecular Weight (g/mol)

225.156

PubChem CID

74776

Formula Weight

225.16

Percent Purity

98%

Chemical Name or Material

mono-Methyl 5-nitroisophthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.