Learn More
ACROBiosystems ACROBiosystems Monoclonal Anti-DNP antibody, Human IgG1 (N297A) Isotype Control (MALS verified)
Supplier:  ACROBiosystems DNPMB2731MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Antibody;1MG;Human IgG1 (N297A)/kappa;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.;Monoclonal Anti-DNP (Anti-Hapten) antibody, Human IgG1 (N297A) was purified from HEK293 cell culture at ACRObiosystems, and specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. ;Immobilized DNP-Albumin Conjugate at 10 μg/mL (100 μL/well) can bind Monoclonal Anti-DNP antibody, Human IgG1 (N297A) Isotype Control (Cat. No. DNP-MB273) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-210-6625