Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate 96.0+%, TCI America™
SDP

Catalog No. D29661G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

66165-37-5

Molecular Formula

C17H29O4P

Molecular Weight (g/mol)

328.39

MDL Number

MFCD06797098

InChI Key

WBSRIXCTCFFHEF-UHFFFAOYNA-N

Synonym

3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Monoethyl Ester, Ethyl Hydrogen 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate

PubChem CID

92274

IUPAC Name

[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl](ethoxy)phosphinic acid

SMILES

CCOP(O)(=O)CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point

128°C

Color

White

Quantity

1 g

Formula Weight

328.39

Percent Purity

≥96.0% (HPLC,T)

Physical Form

Crystalline Powder

Chemical Name or Material

Monoethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate

TSCA

No

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.