Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethyl 5-Nitroisophthalate 98.0+%, TCI America™
SDP

Catalog No. p-7133929 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 22871-55-2
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00130051
InChI Key HOYHBDFTELAGGG-UHFFFAOYSA-N
Synonym 5-Nitroisophthalic Acid Monoethyl Ester
PubChem CID 89872
IUPAC Name 3-ethoxycarbonyl-5-nitrobenzoic acid
SMILES CCOC(=O)C1=CC(=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 239.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monoethyl 5-Nitroisophthalate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.