Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical MooreBrand™ Protection Gown

Catalog No. 19898695
Click to view available options
:
Universal size
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

For Use With (Application) Impervious barrier protection for caregivers in the emergency room as well as in patient rooms
No. of Pockets None
Gender Unisex
Product Type General Purpose Gown
Size Universal
Material Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Finish Embossed
Latex Free Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.