Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morpholine-4-carbothioamide, 97%, Thermo Scientific™

Catalog No. RB00140EA
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

14294-10-1

Molecular Formula

C5H10N2OS

Molecular Weight (g/mol)

146.21

MDL Number

MFCD00178434

InChI Key

GSLBUBZXFUYMSW-UHFFFAOYSA-N

Synonym

4-morpholinecarbothioamide, 1-morpholinethiocarboximide, 4-morpholinethiocarboxamide, morpholine-4-carbothioic acid amide, aminomorpholin-4-ylmethane-1-thione, thiocarbamoylmorpholine, morpholine-4-carbothioicacidamide, 4-amino thiocarbonyl morpholine

PubChem CID

2393544

IUPAC Name

morpholine-4-carbothioamide

SMILES

NC(=S)N1CCOCC1

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

10 g

Formula Weight

146.21

Percent Purity

97%

Chemical Name or Material

Morpholine-4-carbothioamide

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.