Learn More
Scientific Industries TORBAL FTV100L Motorized Test Stand, with FC20 Force Gauge, Wireless Connectivity, 5 Application Modes, and over 15 Programmable Parameters
Supplier:  Scientific Industries FTV100LFC20
Encompass_Preferred
Includes the TORBAL FC20 4lbF force gauge and all essential application modes: Peak, Cycle, Step, Advanced, and Manual. Fully automated and configurable for any tension or compression force testing application. Engineered with a reinforced column, an ultra-high-performance motor, as well as a heavy-duty cross-head to guarantee uninterrupted performance and continuous testing. Precise speed control from 0.4in to 11.8in per minute and 0.001in travel accuracy. The FTVL has 19.6in of travel and a maximum load capacity of 100lbF. Configuring the test for your application is quick and easy with 200 testing profiles. Our intuitive dial knob control allows for an effortless navigation of menu options and input parameters. Included with the test stand: complete set of attachments, shaft extension, wireless antenna, mounting plate, grip knobs, and connection cables. Wireless connectivity between the test stand and force gauge means no tangled or loose cables.
Catalog No. 50-206-7574
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.