Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Adenosine A1R (NP_000665) VersaClone cDNA

Catalog No. RDC2041 Shop All R&D Systems Products
Click to view available options
:
Host Species: Mouse
This item is not returnable. View return policy
This item is not returnable. View return policy

R&D Systems™ Mouse Adenosine A1 R (NP_000665) VersaClone cDNA is an adenosine receptor that belongs to the G protein-coupled receptor 1 family.

The Mouse Adenosine A1 R (NP_000665) VersaClone cDNA clone contains the complete ORF for Adenosine A1R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Adenosine A1R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene mAdora1
Accession Number NP_001008533
Concentration 10 μg at 0.2 μg/μl
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 994bp
pH 8.5
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene ID (Entrez) 134
Host Species Mouse
Gene Alias Adenosine A1 R, adenosine A1 receptor, ADORA 1, ADORA1, RDC7, RDC7adenosine receptor A1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.