Learn More
R&D Systems™ Mouse alpha 1B-Glycoprotein (NP_001074536) VersaClone cDNA
Shop All R&D Systems ProductsSpecifications
Gene
mA1bg
Accession Number
NP_001074536
Concentration
10 μg at 0.2 μg/μL
Formulation
cDNA is provided in 10 mM Tris-Cl
Insert Type
1552bp
pH
8.5
Content And Storage
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species
Mouse
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.