Learn More
Kamiya Biomedical Mouse Hemoglobin ELISA, 96 tests
Supplier:  Kamiya Biomedical KT399
Encompass_Preferred
Hemoglobin ELISA, Mouse. 96 Tests. For the quantitative determination of hemoglobin in mouse biological samples. Assay Range: 12.5 - 400 ng/mL. Includes calibrator and control.
Catalog No. NC0044487
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.