Learn More
ACROBiosystems Mouse IgG2a Kappa Isotype Control (mAb)
Supplier:  ACROBiosystems DNPM487100UG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Mouse IgG2a Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293), which combines the variable region of a mouse monoclonal antibody with Mouse IgG2a constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP.
Catalog No. 50-233-1799
missing translation for 'QuantityLeft'