Learn More
Kingfisher Biotech Inc Mouse IL-36 RA - 500ug
SDP
Supplier:  Kingfisher Biotech Inc RP1999M500
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The Mouse IL-36 Receptor Antagonist yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse IL-36 Receptor Antagonist applications are for cell culture, ELISA standard, and Western Blot Control. Mouse IL-36 Receptor Antagonist yeast-derived recombinant protein can be purchased in multiple sizes. Mouse IL-36 Receptor Antagonist Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD (154)) (Gene ID: 54450). For research use only. Made in the USA
Catalog No. 50-253-2545
missing translation for 'QuantityLeft'