Learn More
R&D Systems™ Mouse Integrin alpha E/CD103 (NP_032425) VersaClone cDNA
Shop All R&D Systems ProductsSpecifications
Product Type
cDNA
Quantity
10μg
Species
Mouse
Gene
mItgae
Accession Number
NP_032425
Concentration
10μg at 0.2μg/μL
Formulation
cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type
3517bp
Storage Requirements
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene ID (Entrez)
3682 (Human); 16407 (Mouse); 83577 (Rat)
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.