Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse P2Y12/P2RY12 (NP_081847.3) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
10 μg
Specifications
Specifications
Gene | P2Y12/P2RY12 |
Accession Number | NP_081847.3 |
Concentration | 10 μg (at 0.2 μg/μL) |
Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
Insert Type | 1057 bp |
Gene ID (Entrez) | 64805 |
Gene Alias | ADPG-R, Gi-coupled ADP receptor HORK3, HORK3, HORK3P2Y12 platelet ADP receptor, P2RY12, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999, SP1999G-protein coupled receptor SP1999 |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Mouse |
Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction