Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Features low endotoxin, low level of lipids, fatty acids and hormones, e.g. insulin, T3, T4.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- Free Fatty Acids ≤0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Insulin, T3, T4
Specifications
Specifications
For Use With (Application) | Mammalian Cell Culture |
Molecular Weight (g/mol) | 66 |
Name | CellMaxx™ Bovine Albumin |
pH Range | 6.5 to 7.5 |
Purification Method | Chromatography |
Quantity | 10 g |
Source | Bovine |
Storage Requirements | 4°C |
Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction