Learn More
MP Biomedicals™ Complement, Rabbit Purified
Shop All MP Biomedicals, Inc ProductsSpecifications
For Use With (Application)
Immunoassay
Name
Rabbit Complement
Quantity
1 mL
Source
Rabbit
Storage Requirements
-20°C
Cross Reactivity
Rabbit
Form
Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.