Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lard

Commonly used in preparation of research animal test diets.

Supplier:  MP Biomedicals™ 0290214005

 View more versions of this product

Catalog No. ICN90214005


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

Use:

Commonly used in the preparation of research animal test diets

Source:

Porcine

Physical Description:

White to cream colored, oily semi-solid to solid

Specifications

Specifications

Animal Diet
Lard is commonly used in the preparation of research animal test diets
Rat
Solid/Semisolid - Greasy or Creamy
2.26 kg
15°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.