Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Lard
Commonly used in preparation of research animal test diets.
Supplier: MP Biomedicals™ 0290214025
Description
Use:
Commonly used in the preparation of research animal test diets
Source:
Porcine
Physical Description:
White to cream colored, oily semi-solid to solid
Specifications
Animal Diet | |
Lard is commonly used in the preparation of research animal test diets | |
Rat |
Solid/Semisolid - Greasy or Creamy | |
11.33 kg | |
15°C to 30°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction