Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Malt Extract Agar
Shop All MP Biomedicals, Inc Products
Click to view available options
:
500g
Specifications
Specifications
Content And Storage | 15°C to 30°C |
Form | Powder |
Media Type | Malt Extract Agar |
Product Type | Agar |
Formulation | Malt extract 12.75g/L, Dextrin 2.75g/L, Glycerin 2.35g/L, Gelatin Peptone 0.78g/L, Agar 15g/L, 4.6 ±0.2 final pH |
For Use With (Application) | Cultivation of and enumeration of Yeasts and filamentous fungi (mold) from cosmetics and other samples |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction