Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Sephadex™ G-100

Catalog No. ICN19525510 Shop All MP Biomedicals, Inc Products
Click to view available options
:

Fractionation Range (MW)

  • Dry Bead Diameter: 40 to 120μm
  • CAS: 9050-94-6

Specifications

Quantity

10g

Flow Rate

≥40 cm/hr.

Particle Size

40 to 120 μm

Bed Volume

15 to 20mL/g in Distilled Water

Product Type

Sephadex G-100 Media

Format

Powder

For Use With (Application)

Desalting and group separations of low and high molecular weight molecules, Buffer exchange and the removal of small molecules during the preparation of large biomolecules, such as ampholytes, detergents

Max. Pressure

0.96 bar

Support Type

Cross-linked Dextran with Epichlorohydrin

Sample Type

Proteins, Enzymes and Peptides

Content And Storage

15° to 30°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.