Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SOB, Powder

Catalog No. MP113030022 Shop All MP Biomedicals, Inc Products
Click to view available options
:

Standard media for preparing cells for chemical transformation

  • Contents/liter: 20 g tryptone, 5 g yeast extract, 0.5 g NaCl, 5 g MgSO4·7H2O
  • Use 31 g powder per liter

Specifications

Content And Storage

15°C to 30°C

Form

Powder

Product Type

Microbiology Media

Formulation

Tryptone 20g/L, Yeast extract 5g/L, NaCl 0.5g/L, MgSO4.7H2O 5g/L

Quantity

454 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.