Learn More
Molecular Dimensions MRC PLATE 96 WELL 2 DROP (40)
MRC PLATE 96 WELL 2 DROP (40) Polystyrene Designed by crystallographers at the MRC in Cambridge. Excellent optical properties.Raised, wide wells make crystal retrieval easy.The wells are conical. 10nl-5ul drops and the reservoirs hold volumes from 50-100 ul.