Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRS 1334, Tocris Bioscience™

Catalog No. p-4785141 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, highly selective hA3 antagonist

Potent and highly selective antagonist for the human adenosine A3 receptor. Ki values are 2.69 nM at hA3, and > 100 μM at rat A1 and rat A2A receptors.

Chemical Identifiers

CAS 192053-05-7
Molecular Formula C31H26N2O6
Molecular Weight (g/mol) 522.557
InChI Key WICWQHSRWZIPNB-AREMUKBSSA-N
Synonym 1, 4-Dihydro-2-methyl-6-phenyl-4-(phenylethynyl)-3, 5-pyridinedicarboxylic acid 3-ethyl-5-[(3-nitrophenyl)methyl] ester
PubChem CID 98072178
IUPAC Name 3-O-ethyl 5-O-[(3-nitrophenyl)methyl] (4R)-2-methyl-6-phenyl-4-(2-phenylethynyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C#CC2=CC=CC=C2)C(=O)OCC3=CC(=CC=C3)[N+](=O)[O-])C4=CC=CC=C4)C

Specifications

Quantity 10 mg
Formula Weight 522.56
Percent Purity >98%
Chemical Name or Material MRS 1334
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.