Learn More
TARGETMOL CHEMICALS INC MRS 1754 10MG
Supplier:  TARGETMOL CHEMICALS INC T16140
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MRS 1754 is a selective antagonist radioligand for the A2B adenosine receptor. It has a very low affinity for A1 and A3 receptors of both humans and rats. purity: 97%
Catalog No. 50-258-1189
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.