Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRS 1754, Tocris Bioscience™

Catalog No. p-4784167 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective A2B antagonist

Selective adenosine A2B receptor antagonist (Ki values are 1.97, 16.8, 403, 503, 570 and 612 nM for hA2B, rA1, hA1, hA2A, hA3 and rA2A receptors respectively).

Chemical Identifiers

CAS 264622-58-4
Molecular Formula C26H26N6O4
Molecular Weight (g/mol) 486.532
InChI Key AJBBEYXFRYFVNM-UHFFFAOYSA-N
Synonym n-4-cyanophenyl-2-4-2,6-dioxo-1,3-dipropyl-7h-purin-8-yl phenoxy acetamide, n-4-cyanophenyl-2-4-2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1h-purin-8-yl phenoxy-acetamide, lopac-m-6316, d01qyq, d0ne5p, atl-gw-8, gtpl449, gtpl453, atl-gw-22, a2b adenosine receptor antagonists, adenosine/ortho-mcneil
PubChem CID 6603931
IUPAC Name N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetamide
SMILES CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=C(C=C3)OCC(=O)NC4=CC=C(C=C4)C#N

Specifications

Quantity 10 mg
Formula Weight 486.52
Percent Purity >98%
Chemical Name or Material MRS 1754
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.