Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ Pro Single Gas Detector

Provides worry-free performance, standing up to the roughest handling in tough industrial environments

Specifications

Components Measured Hydrogen Sulfide
Type Single Gas Detector
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
19-810-243
View Documents
MSA™
10096702
Single Pack N/A N/A N/A
19-815-621
View Documents
MSA™
10096703
Bulk Pack N/A N/A N/A
Description

Description

  • Built-in gas delivery adapter; no accessories are needed for gas response tests
  • Distinctive triple alarm system leaves no doubt for users in alarm situations
  • Extended frequency range covers a wide sound spectrum
  • Visual alarm features dual LED displays located on the unit's top, visible from all angles
  • Internal vibrating alarm is standard, emitting a pulse that directly calls additional attention when in alarm mode

Detects Hydrogen Sulfide

Certifications

Class I, Div. 1, Groups A, B, C and D; Class II, Div. 1, Group G, AEx ia IIC T4

Warranty and Services

Two Years from date of purchase

Specifications

Specifications

Hydrogen Sulfide
Single Gas Detector
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.