Learn More
MSA™ Air-Line Respirator / Air-Supplied Hood Accessory, Quick-disconnect Male Plug with Male NPT
For consistent reliability when connecting an inlet pressure gauge, choose an MSA brass or aluminum Quick-disconnect for your system.
$105.27
Specifications
Connection Type | Male 0.25 in. NPT |
---|---|
For Use With (Equipment) | Airline Respirator and Air-supplied Hood |
Catalog Number | Mfr. No. | Material | dimType | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | dimType | Price | Quantity & Availability | ||||
18-139-737
|
MSA™
473501 |
Brass |
Each of 1 for $105.27
|
|
|||||
Description
Male 1/4 in. NPT
Fittings with the same letter code are interchangeable.
- AL: Aluminum
- S: Steel
- SST: Stainless Steel
- BR: Brass
Specifications
Male 0.25 in. NPT | |
Airline Respirator and Air-supplied Hood |