Learn More
Liofilchem™ MTS™ Cefotetan [CTT] 0.016-256 µg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
Supplier: Liofilchem™ 920200
Description
Specifications
Cefotetan | |
-20°C or 2°C to 8°C until the given expiry date | |
CTT | |
Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms | |
100/Pk. |
0.016-256 μg/mL | |
Liofilchem™ MTS™ Cefotetan | |
MIC Test Strip | |
Canister Pack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.