Learn More
Gold Biotechnology Inc MUG, 1 g
SDP
Supplier:  Gold Biotechnology Inc MUG1
Encompass_Preferred
MUG14-Methylumbelliferyl beta-D-glucuronide (MUG) is a fluorogenic substrate for beta-glucuronidase (GUS). Upon cleavage, MUG yields blue fluorescence seen under UV light.This substrate is commonly used for the detection of Escherichia coli through its ability to detect activity of beta-glucuronidase, which is encoded by uidA. GUS activity is tested in minimal medium o-nitrophenyl-beta-D-galactopyranoside (MMO)-MUG.It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.
Catalog No. 50-153-2816
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.