Learn More
Hoefer, Inc Hoefer™ Multiple Gel Casters for SE250&SE260 Mighty Small Mini Vertical Electrophoresis Unit
Supplier: Hoefer, Inc SE235
Enable several gradient or homogenous gels to be cast simultaneouslyMultiple Gel Casters• Pour uniform gels from the top; pump gradient gels into an inlet port at the bottom of the unit• Sandwiches fill at the same time at the same rate from the same solution.The result: identical gels.• Glass plate sandwiches seal leak free without the use of messy grease or tape• Once cast, gels can be wrapped in plastic and refrigerated for several weeksSE215: Ten Gel Caster for 10 x 8 cm platesIncludes: 10 x 8 cm Rectangular Glass Plates–20 pcs10 x 8 cm Notched Alumina Plates–10 pcsSpring Clamps–2 pcsSE275:Four Gel Caster for 10 x 8 cm platesIncludes: 10 x 8 cm Rectangular Glass Plates–10 pcs10 x 8 cm Notched Alumina Plates–4 pcsSpring Clamps–2 pcsSE235:Four Gel Caster for 10 x 10.5 cm platesIncludes: 10 x 10.5 cm Rectangular Glass Plates–5 pcs10 x 10.5 cm Notched Alumina Plates–4 pcsSpring Clamps–4 pcs
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.