Learn More
Beckman Coulter Multisizer™ 4e Smart Technology Aperture Tubes
Smart Technology Aperture Tubes are designed specifically for the Multisizer 4 or 4e Analyzer. The aperture allows extended sizing measurement range of 2% to 80% of the aperture diameter.
Supplier: Beckman Coulter A36392
Description
- Extended measurement range (2% to 80% of aperture diameter) leads to less frequent aperture changes for more analysis options
- A wide range of apertures is available with diameters ranging from 10 to 2,000 μm
- Specifically designed for the Multisizer 4e or Multisizer 4 Coulter Counters
- 100 μm ST Aperture Tube is standard with the Multisizer 4e Coulter Counter
- Compatible with ST Beakers and Accuvette STs
- Convenient tubes ensure accurate, and consistent results
- Size and serial number information is included on a bar code on the tube for automatic data entry via instrument bar code reader
Specifications
Aperature Tube | |
1.0 μm to 40 μm | |
Multisizer™ 4 or 4e Coulter Counters |
Glass | |
50 μm |