Learn More
Enzo Life Sciences MULTIVIEW (mouse-HRP/rabbit-AP) IHC kit (1x100tests)
SDP
Supplier:  Enzo Life Sciences ADI9501000001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Superior multiplex IHC kit for efficient detection of multiple antigens in a single tissue sample. The MULTIVIEW (mouse-HRP/rabbit-AP) IHC kit is a non-biotin one-step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and cryostat sections. The kit may also be used with blood smears, cytosmears, and cell preparations. Assay Time: ~2 hours. Applications: IHC. Contents: Peroxidase Block, POLYVIEW IHC reagent (mouse-HRP), POLYVIEW IHC reagent (rabbit-AP), HIGHDEF IHC chromogen substrate (DAB, stable) buffer & chromogen, HIGHDEF red IHC chromogen (AP) buffer & chromogen, Empty mixing bottles
Catalog No. NC0996478
missing translation for 'QuantityLeft'