Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscimol, Tocris Bioscience™

Catalog No. p-4785157 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent GABAA agonist; also GABAA-ρ partial agonist

Potent GABAA receptor agonist and partial GABAA-ρ receptor agonist. Inhibits memory retention and attenuates airway constriction in vivo.

Chemical Identifiers

CAS 2763-96-4
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00057894
InChI Key ZJQHPWUVQPJPQT-UHFFFAOYSA-N
Synonym muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole
PubChem CID 4266
ChEBI CHEBI:7035
IUPAC Name 5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one
SMILES NCC1=CC(=O)NO1

Specifications

Quantity 1 mg
Formula Weight 114.1
Percent Purity >98%
Chemical Name or Material Muscimol
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.