Learn More
Sigma Aldrich Fine Chemicals Biosciences Mutanolysin from Streptomyces globisporus ATCC 21553, 55466-22-3, MFCD00131635, 10KU
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M990110KU
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Lyophilized powder, ≥4000 units/mg protein (biuret), Chromatographically purified. Mutanolysin from Streptomyces globisporus consists of two main lytic enzymes and may be a useful agent for dental caries control.
Catalog No. NC1037321
missing translation for 'QuantityLeft'