Learn More
Sigma Aldrich Fine Chemicals Biosciences N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 1760-24-3, MFCD00008173, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 104884100ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Linear Formula: (CH3O)3Si(CH2)3NHCH2CH2NH2, Molecular Weight: 222.36, 97%, Synonym: N-(2-Aminoethyl)-3-(trimethoxysilyl)propylamine, [3-(2-Aminoethylamino)propyl]trimethoxysilane. Has been used as an organic ligand for the surface modification of silica gel to uptake heavy metal ions. It can react with fluorinated carbon nanotubes (F-CNT) to form aminoalkylalkoxysilane functionalized carbon nanotubes.
Catalog No. NC0956250