Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-tert-butylbenzyl)-N-methylamine, Thermo Scientific™

Catalog No. MO08598DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 65542-26-9
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
InChI Key ZSHCHOYJMLEAOX-UHFFFAOYSA-N
Synonym n-4-tert-butylbenzyl-n-methylamine, 4-tert-butylphenyl methyl methyl amine, 1-4-tert-butyl phenyl-n-methylmethanamine, 1-4-tert-butylphenyl-n-methylmethanamine, 4-tert-butylbenzylmethylamine, n-methyl-4-tert-butylbenzylamine, n-methyl-4-tert.butylbenzylamine, n-methyl 4-tert-butylbenzyl amine, 4-tert-butyl phenyl methyl methylamine, benzenemethanamine,4-1,1-dimethylethyl-n-methyl
PubChem CID 2396874
IUPAC Name 1-(4-tert-butylphenyl)-N-methylmethanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CNC

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.29
Percent Purity 97%
Chemical Name or Material N-(4-tert-butylbenzyl)-N-methylamine
Hazard Category GHS Signal Word: Danger
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.